KN-93 phosphate ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: (E)-N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide phosphate, (E)-KN-93 phosphate

AG-CR1-0065-M001 AG-CR1-0065-M005 AG-CR1-0065-M025
102516-108EA 34.62 USD
102516-108 102516-110 102516-112
KN-93 phosphate ≥98%
KN-93 phosphate

Selective Ca2+/calmodulin-dependent protein kinase II inhibitor. Insulin release inhibitor. Acid secretion inhibitor. G1 cell cycle arrest inducer. Apoptosis inducer. Ion channel inhibitor. ROS inducer. Androgen receptor inhibitor.

Formula: C₂₆H₂₉ClN₂O₄S·H₃PO₄
MW: 501.0·98.0 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03788201
CAS Number: 1188890-41-6 | 139298-40-1 (free base)

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR