CHS-828 ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 1-[6-(4-Chlorophenoxy)hexyl]-3-cyano-2-pyridin-4-ylguanidine

AG-CR1-0064-M005 AG-CR1-0064-M025
102516-104EA 83.08 USD
102516-104 102516-106
CHS-828 ≥98%
CHS-828

Antitumor compound. Programmed cell death inducer. Cytotoxic. Antiproliferative. p53 activator. IkappaB kinase inhibitor. NF-kappaB inhibitor. Nampt/visfatin inhibitor.

Formula: C₁₉H₂₂ClN₅O
MW: 371.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03700766
CAS Number: 200484-11-3

Order Now

Specification Test Results

Purity 98% (HPLC & NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, 100% ethanol or dimethylformamide.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR