Necrostatin-7 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 5-((3-(4-Fluorophenyl)-1H-pyrazol-4-yl)methylene)-2-imino-3-(thiazol-2-yl)thiazolidin-4-one

AG-CR1-0054-M005 AG-CR1-0054-M025
102516-084EA 110.77 USD
102516-084 102516-086
Necrostatin-7 ≥98% (by NMR)
Necrostatin-7

Necroptosis inhibitor. Does not inhibit RIP1 kinase.

Formula: C₁₆H₁₀FN₅OS₂
MW: 371.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02334728
CAS Number: 351062-08-3

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO or 100% ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR