Necrostatin-5 ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CR1-0049-M005
AG-CR1-0049-M025
102516-068EA
55.38
USD
102516-068
102516-070
Necrostatin-5 ≥98% (by NMR)
Necrostatin-5
Necroptosis inhibitor. Indirect RIP1 kinase inhibitor. Cardioprotective
Formula:
C₁₉H₁₇N₃O₂S₂ Storage Temperature: Refrigerator |
CAS Number:
337349-54-9 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility |
Soluble in DMSO ( |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...