bpV(bipy) ≥98%

Supplier: ADIPOGEN CORP MS

AG-CR1-0047-M005 AG-CR1-0047-M025
102516-064EA 96.92 USD
102516-064 102516-066
bpV(bipy) ≥98%
bpV(bipy) hydrate

Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic in vitro and in vivo. PTEN inhibitor.

Formula: K[VO(O₂)₂C₁₀H₈N₂]·3H₂O
MW: 326.2·54.0 g/mol
Storage Temperature: Freezer
CAS Number: 1173697-60-3 | 42494-71-3 (anhydrous)

Order Now

Specification Test Results

Purity 98% (51V-NMR)
Appearance/Color Yellow to orange crystalline solid.
Solubility Soluble in DMSO or water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR