bpV(pic) ≥96% (by 51V-NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-0043-M005 AG-CR1-0043-M025
102516-048EA 83.08 USD
102516-048 102516-050
bpV(pic) ≥96% (by 51V-NMR)
bpV(pic)

Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic. Angiogenesis inhibitor. PTEN inhibitor.

Formula: K₂[VO(O₂)₂C₆H₄NO₂]·2H₂O
MW: 331.2·36.0 g/mol
Storage Temperature: Freezer
CAS Number: 148556-27-8

Order Now

Specification Test Results

Purity 96% (51V-NMR)
Appearance/Color Yellow crystalline solid.
Solubility Soluble in DMSO or water. NOTE: The product is not stable in water. Decomposition starts immediately after solubilization. Rate of decomposition is slower in dry DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR