(-)-Indolactam V ≥97% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0041-C300 AG-CR1-0041-M001
102516-040EA 263.08 USD
102516-040 102516-042
(-)-Indolactam V ≥97% (by NMR)
(-)-Indolactam V

Tumor promoter. Protein kinase C (PKC) inducer. Mitogenesis activator. Differentiation inducer.

Formula: C₁₇H₂₃N₃O₂
MW: 301.39 g/mol
Storage Temperature: Freezer
CAS Number: 90365-57-4

Order Now

Specification Test Results

Purity 97% (NMR)
Appearance/Color Off-white to light yellow solid.
Solubility Soluble in DMSO or 100% ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR