(S)-CR8 98

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0040-M001 AG-CR1-0040-M005
102516-036EA 83.08 USD
102516-036 102516-038
(S)-CR8 98
(S)-CR8

Potent and selective cyclin-dependent kinase (CDK) 1, 2, 5, 7 and 9 inhibitor. Apoptosis inducer. Glycogen synthase kinase (GSK-3alpha/beta) inhibitor.

Formula: C₂₄H₂₉N₇O
MW: 431.5 g/mol
Storage Temperature: Freezer
CAS Number: 1084893-56-0

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR