(R)-CR8 98

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (R)-2-((9-Isopropyl-6-((4-(pyridin-2-yl)benzyl)amino)-9H-purin-2-yl)amino)butan-1-ol

AG-CR1-0039-M001 AG-CR1-0039-M005
102516-032EA 83.08 USD
102516-032 102516-034
(R)-CR8 98
(R)-CR8

Potent and selective cyclin-dependent kinase (CDK) 1, 2, 5, 7 and 9 inhibitor. Apoptosis inducer. Potent glycogen synthase kinase (GSK-3alpha/beta) inhibitor.

Formula: C₂₄H₂₉N₇O
MW: 431.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18785641
CAS Number: 294646-77-8

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR