Alsterpaullone ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 7,12-Dihydro-9-nitroindolo[3,2-d][1]benzazepin-6(5H)-one

AG-CR1-0036-M001 AG-CR1-0036-M005
102516-020EA 55.38 USD
102516-020 102516-022
Alsterpaullone ≥98%
Alsterpaullone

Potent CDK1/cyclin B (IC50 = 35 nM) inhibitor. Anti-tumor compound Potent inhibitor of CDK2/cyclin A, CDK2/cyclin E (IC50 = 200 nM), CDK5/p25 (IC50 = 40 nM), CDK5/p35 (IC50 = 40 nM). GSK-3beta (glycogen synthase kinase-3beta) inhibitor. Apoptosis inhibitor. Apoptosis inducer. Angiogenesis inhibitor.

Formula: C₁₆H₁₁N₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD02683579
CAS Number: 237430-03-4

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Yellow to brown powder.
Solubility Soluble in DMSO; insoluble in water or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR