1-Azakenpaullone ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 9-Bromo-7,12-dihydropyrido[3',2':2,3]azepino[4,5-b]indol-6(5H)-one

AG-CR1-0035-M001 AG-CR1-0035-M005
102516-016EA 103.85 USD
102516-016 102516-018
1-Azakenpaullone ≥98%
1-Azakenpaullone

Potent and ATP-competitive GSK-3beta (glycogen synthase kinase-3beta) inhibitor (IC50 = 18 nM).

Formula: C₁₅H₁₀BrN₃O
MW: 328.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09037522
CAS Number: 676596-65-9

Order Now

Specification Test Results

Purity 98% (HPLC, NMR)
Appearance/Color Off-white to tan solid.
Solubility Soluble in DMSO; insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR