SB203580 ≥98%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
SB 203580, 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine
AG-CR1-0030-M001
AG-CR1-0030-M005
AG-CR1-0030-M025
102515-988EA
41.54
USD
102515-988
102515-990
102515-992
SB203580 ≥98%
SB203580
Cell permeable, specific p38 MAP kinase inhibitor. Binds to the ATP binding site of p38 MAP kinase. T cell proliferation inhibitor. IL-2 production inhibitor. SAPK/JNK inhibitor. COX-1 and -2 inhibitor. Raf-1 activator. Apoptosis enhancer. Antiproliferative. PDK1 inhibitor. Anti-inflammatory. Review. Enhances cardiomyogenesis of human embryonic stem cells (hESCs).
Formula:
C₂₁H₁₆FN₃OS MW: 377.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00922198 CAS Number: 152121-47-6 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...