MnTMPyP·pentachloride ≥96%
Supplier: ADIPOGEN CORP MS
AG-CR1-0026-M010
AG-CR1-0026-M050
102515-970EA
34.62
USD
102515-970
102515-972
MnTMPyP·pentachloride ≥96%
MnTMPyP pentachloride
Cell permeable superoxide dismutase (SOD) mimetic. Peroxynitrite scavenger. Displays a protective effect against H2O2 mediated injury. Neuroprotective. Oxidative stress inhibitor. Apoptosis inhibitor. Reduces inflammation.
Formula:
C₄₄H₃₆MnN₈Cl₅ MW: 909.0 g/mol Storage Temperature: Freezer |
CAS Number:
100012-18-8 |
Specification Test Results
Purity |
|
Appearance/Color | Black solid. |
Solubility | Soluble in aqueous base. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...