1-Methyl-3,4-bis(3-indolyl)maleimide ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
3,4-Di(1H-indol-3-yl)-1-methyl-1H-pyrrole-2,5-dione, Bisindolylmaleimide V
AG-CR1-0023-M001
AG-CR1-0023-M005
102515-954EA
76.15
USD
102515-954
102515-956
1-Methyl-3,4-bis(3-indolyl)maleimide ≥98% (by NMR)
1-Methyl-3,4-bis(3-indolyl)maleimide
Negative control for protein kinase C (PKC) inhibitors. S6K inhibitor. Necrosis inhibitor.
Formula:
C₂₁H₁₅N₃O₂ MW: 341.37 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00236433 CAS Number: 113963-68-1 |
Specification Test Results
Purity |
|
Appearance/Color | Red crystalline solid. |
Solubility | Soluble in DMSO or methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...