1400W dihydrochloride ≥98%
Supplier: ADIPOGEN CORP MS
Synonyms:
1400W 2HCl, N-(3-(Aminomethyl)benzyl)acetimidamide dihydrochloride
AG-CR1-0018-M005
AG-CR1-0018-M025
AG-CR1-0018-M100
102515-928EA
55.38
USD
102515-928
102515-930
102515-932
1400W dihydrochloride ≥98%
1400W dihydrochloride
A slow, tight binding and highly selective inhibitor of iNOS (inducible nitric oxide synthase/NOS II) (Kd
Formula:
C₁₀H₁₅N₃·2HCl MW: 250.167 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03428622 CAS Number: 214358-33-5 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in water, methanol or DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...