BAY 11-7082 ≥99% (by NMR)
Supplier: ADIPOGEN CORP MS
AG-CR1-0013-M010
AG-CR1-0013-M050
102515-910EA
96.92
USD
102515-910
102515-912
BAY 11-7082 ≥99% (by NMR)
BAY 11-7082
IkappaBalpha kinase inhibitor. NF-kappaB inhibitor. Potential anti-inflammatory agent. Apoptosis inducer. Inhibits the release of proinflammatory cytokines. NLRP3 inflammasome inhibitor. Reduces ATPase activity of the NLRP3 inflammasome. Inhibits platelet aggregation.
Formula:
C₁₀H₉NO₂S Storage Temperature: Freezer |
MDL Number:
MFCD00712162 CAS Number: 19542-67-7 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white crystalline solid. |
Solubility | Soluble in DMSO, 100% ethanol, dichloromethane or ethyl acetate; insoluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...