Bisindolylmaleimide II ≥97%

Supplier: ADIPOGEN CORP MS

AG-CR1-0010-M001 AG-CR1-0010-M005
102515-902EA 76.15 USD
102515-902 102515-904
Bisindolylmaleimide II ≥97%
Bisindolylmaleimide II

Protein kinase C (PKC) inhibitor. Binds with reversed orientation to protein kinase A (PKA).

Formula: C₂₇H₂₆N₄O₂
MW: 438.5 g/mol
Storage Temperature: Freezer
CAS Number: 137592-45-1

Order Now

Specification Test Results

Purity 97% (mixture of 2 isomers) (NMR)
Appearance/Color Orange solid.
Solubility Soluble in DMSO or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR