(R)-Roscovitine ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine, Roscovitine, (R)-2-((6-(Benzylamino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol

AG-CR1-0006-M001 AG-CR1-0006-M005 AG-CR1-0006-M050
102515-884EA 41.54 USD
102515-884 102515-886 102515-888
(R)-Roscovitine ≥98% (by NMR)
(R)-Roscovitine

Potent and selective inhibitor of CDKs. More potent than olomoucine. Inhibits CDK1/cyclin B kinase (IC50 = 450 nM), CDK2 (IC50 = 700 nM) and CDK5/p35 (IC50 = 160 nM). Inhibits M phase promoting factor (MPF) kinase activity. Arrests human fibroblasts in G1 phase. Antitumor compound. Activates the mitogen-activated protein kinase pathway. Targets both the p53 and NF-kappaB pathways[10]. Has effects on calcium channel gating. Prevents DNA damage-induced cyclin A1 upregulation. Apoptosis inducer. As CYC202 in phase I clinical trials. Reviews.

Formula: C₁₉H₂₆N₆O
MW: 354.45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02266401
CAS Number: 186692-46-6

Order Now

Specification Test Results

Purity >98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or 100% ethanol; only moderately soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR