Pifithrin-alpha·hydrobromide ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
AG-CR1-0004-M005
AG-CR1-0004-M010
AG-CR1-0004-M025
102515-874EA
69.23
USD
102515-874
102515-876
102515-878
Pifithrin-alpha·hydrobromide ≥98% (by NMR)
Pifithrin-alpha hydrobromide
p53 inhibitor. Reversibly blocks p53-dependent transcriptional activation. Tool to combat side effects of cancer therapy. Apoptosis inhibitor.
Formula:
C₁₆H₁₈N₂OS·HBr Storage Temperature: Freezer |
MDL Number:
MFCD00417851 CAS Number: 63208-82-2 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO or 100% ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...