N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-0003-M005 AG-CR1-0003-M025
102515-870EA 69.23 USD
102515-870 102515-872
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride ≥98% (by NMR)
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride

Calmodulin antagonist. Cell proliferation inhibitor. Ca2+/calmodulin-regulated phosphodiesterase (PDE I) inhibitor. Myosin light chain kinase (MLCK) inhibitor.

Formula: C₁₆H₂₂N₂O₂S
Storage Temperature: Freezer
MDL Number: MFCD00058043
CAS Number: 61714-25-8

Order Now

Specification Test Results

Purity >98% (NMR)
Appearance/Color White to off-white crystalline solid.
Solubility Soluble in DMSO or 100% ethanol (slightly soluble).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR