H-89 dihydrochloride ≥98%

Supplier: ADIPOGEN CORP MS

AG-CR1-0002-M001 AG-CR1-0002-M005 AG-CR1-0002-M025
102515-864EA 34.62 USD
102515-864 102515-866 102515-868
H-89 dihydrochloride ≥98%
H-89 dihydrochloride

Cell permeable potent and selective cAMP- and cGMP-dependent protein kinase (PKA and PKG) inhibitor. Protein kinase Cµ (PKCµ) inhibitor. Ca2+/calmodulin-dependent protein kinase II inhibitor. Casein kinase I inhibitor. Myosin light chain kinase (MLCK) inhibitor. Apoptosis enhancer. Tool to study protein crystal structure-inhibitor interactions. Rho kinase inhibitor. Cell proliferation inhibitor. Review.

Formula: C₂₀H₂₀BrN₃O₂S·2HCl
Storage Temperature: Refrigerator
CAS Number: 127243-85-0

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR