H-8·dihydrochloride ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-0001-M010 AG-CR1-0001-M050
102515-860EA 55.38 USD
102515-860 102515-862
H-8·dihydrochloride ≥98% (by NMR)
H-8 dihydrochloride

Potent cAMP- and cGMP-dependent protein kinase (PKA and PKG) inhibitor. Myosin light chain kinase (MLCK) inhibitor. Tool to study protein crystal structure-inhibitor interactions.

Formula: C₁₂H₁₅N₃O₂S·2HCl
MW: 265.3·72.9 g/mol
Storage Temperature: Refrigerator
CAS Number: 113276-94-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or 100% ethanol (slightly soluble).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR