Resveratrol

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

AG-CN2-0033-M050 AG-CN2-0033-M100 AG-CN2-0033-M500
102515-790EA 34.62 USD
102515-790 102515-792 102515-794
Resveratrol
Resveratrol

Potent phenolic antioxidant found in grapes and red wine. Eicosanoid synthesis and platelet aggregation inhibitor. Estrogen receptor agonist. Chemopreventive. Specific inhibitor of cyclooxygenase-1 (COX-1). Anti-inflammatory. Ribonucleotide reductase and DNA synthesis. Arrests cell cycle at S/G2 phase. Anticancer and antiproliferative compound. Apoptosis inducer. Protein kinase D inhibitor. Does not inhibit PKC. Autophagy inducer. Potent SIRT1 (sirtuin 1) activator. Neuroprotective. Adipogenesis inhibitor. PGC-1alpha activator. Sonic hedgehog (Shh) signaling pathway modulator. Gli1 mRNA expression inhibitor. Downregulates Gli transcriptional activity. Cardioprotective. Anti-diabetic. Senescence modulator. Extends lifespan. Inhibitor of NLRP3 inflammasome activation.

Formula: C₁₄H₁₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00133799
CAS Number: 501-36-0

Order Now

Specification Test Results

Source/Host Isolated from Polygonum cuspidatum.
Purity 98% (HPLC)
Appearance/Color Off-white to greyish powder.
Solubility Soluble in DMSO, ethanol or DMF. Slightly soluble in water or PBS.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR