Piperlongumine ≥97%
Supplier: ADIPOGEN CORP MS
Synonyms:
(E)-1-(3-(3,4,5-Trimethoxyphenyl)acryloyl)-5,6-dihydropyridin-2(1H)-one
Cytotoxic against tumor cell lines. Induces necrosis and apoptosis in cancer cells. Shows anti-platelet aggregation activity possibly by inhibition of cyclooxgenase activity and a decrease in thromboxane A2 formation. Shows significant anxiolytic and antidepressant activities. Promotes adipogenesis of 3T3-L1 cells. Induces in vivo and in vitro mutagenicity in eukaryotic models. Selectively kills cancer cells by targeting the stress response to ROS. Shows in vitro schistosomicidal activity.
Formula:
C₁₇H₁₉NO₅ MW: 317.341 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00075706 CAS Number: 20069-09-4 |
Specification Test Results
Source/Host | Isolated from Piper longum roots. |
Purity |
|
Appearance/Color | White crystals. |
Solubility | Soluble in DMSO, ethanol, methanol and dimethylformamide. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...