Radicicol (from Gilmaniella species) ≥99% (by HPLC)

Supplier: ADIPOGEN CORP MS

AG-CN2-0021-M001 AG-CN2-0021-M005
102515-734EA 55.38 USD
102515-734 102515-736
Radicicol (from Gilmaniella species) ≥99% (by HPLC)
Radicicol

Radicicol is an antibiotic, protein tyrosine kinase inhibitor.


  • Cyclooxygenase-2 (COX-2) expression inhibitor
  • Suppresses expression of iNOS (NOS II)
  • Apoptosis inducer


Antibiotic. Protein tyrosine kinase inhibitor. Has anti-fungal, anti-malarial, anti-angiogenic (in vivo), anti-inflammatory and anti-viral activity. Cyclooxygenase-2 (COX-2) expression inhibitor without affecting COX-1 expression in LPS-stimulated macrophages. Induces the differentiation of HL-60 cells into macrophages. Potent HSP90 inhibitor. Anticancer compound. In vivo Rad/Raf interaction inhibitor. Inhibitor of AP-1-, NF-kappaB- and serum response factor (SRF)-mediated transcription. Non-competitive inhibitor of ATP citrate lyase. DNA topoisomerase VI and type II DNA topoisomerase inhibitor. Protects against LPS/IFN-gamma-induced neuronal cell death.


White to slightly yellow solid


Stable for at least two years after receipt when stored at -20°C. Store solutions at -20°C in the dark.


Soluble in DMSO, methanol, acetone, ethyl acetate or chloroform.

Formula: C₁₈H₁₇ClO₆
MW: 364.8 g/mol
Storage Temperature: Freezer
CAS Number: 12772-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR