α-Galactosylceramide ≥96% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CN2-0013-C250 AG-CN2-0013-M001
102515-710EA 152.31 USD
102515-710 102515-712
α-Galactosylceramide ≥96% (by NMR)
α-Galactosylceramide

Synthetic. Originally isolated from the marine sponge Angelas mauritianus. Immunostimulant. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells. Protects against LPS-induced shock. Potent antitumor compound.


  • Immunostimulant
  • Potent stimulator of natural killer T (NKT) cells
  • Potent antitumour compound
  • Purity: ≥96% (NMR)


Stability: Soluble in pyridine. Stable for at least 2 years after receipt when stored at long term storage −20 °C. Short term storage at +4 °C.

Formula: C₅₀H₉₉NO₉
Storage Temperature: Freezer
MDL Number: MFCD00939559
CAS Number: 158021-47-7

Order Now

Specification Test Results

Purity >96% (NMR)
Appearance/Color Off-white solid.
Solubility Soluble in pyridine. Solubilization in Tween20 or PBS for cell culture use see online protocol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR