Hyperforin DCHA ≥97%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Hyperforin dicyclohexylammonium salt, (1R,5S,6R,7S)-4-Hydroxy-5-isobutyryl-6-methyl-1,3,7-tris(3-methylbut-2-en-1-yl)-6-(4-methylpent-3-en-1-yl)bicyclo[3.3.1]non-3-ene-2,9-dione dicyclohexylamine

AG-CN2-0008-C500 AG-CN2-0008-M001
102515-684EA 166.15 USD
102515-684 102515-686
Hyperforin DCHA ≥97%
Hyperforin DCHA

Key constituent of St. John's wort. Shows properties of potential pharmacological interest, including antibacterial, anti-malarial, anti-inflammatory, anti-cancer and anti-angiogenic effects. Anti-depressant and anxiolytic compound. Specific activator of TRPC6 channels. Inhibits the re-uptake of neurotransmitters in synapses (serotonin, norepinephrine, dopamine, GABA, glutamate). Activator of the pregnane X receptor (PXR). Regulates expression of the cytochrome P450 CYP3A4 and CYP2C9 and hepatic drugs metabolism. Potential anti-Alzheimer compound. Potent SIRT1 (sirtuin 1) and SIRT2 (sirtuin 2) inhibitor.

Formula: C₃₅H₅₁O₄·C₁₂H₂₄N
MW: 535.8·182.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04000098
CAS Number: 238074-03-8

Order Now

Specification Test Results

Source/Host Isolated from St. John's wort (Hypericum perforatum).
Purity 97% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in 100% ethanol, methanol or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR