Thapsigargin ≥98% (by HPLC), high purity
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-(((Z)-2-methylbut-2-enoyl)oxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate
AG-CN2-0003-M001
AG-CN2-0003-M005
AG-CN2-0003-M010
102515-660EA
96.92
USD
102515-660
102515-662
102515-664
Thapsigargin ≥98% (by HPLC), high purity
Thapsigargin
Intracellular Ca2+ signaling probe. Specific and sensitive inhibitor of SERCA. Non-TPA/PMA tumor promoter. Histamine release inducer. Apoptosis inducer. Mitochondrial dysfunction inducer. NOS modulator. Angiogenesis inhibitor. Stimulator of arachidonic acid metabolism in macrophages. Autophagy inducer. TRAIL sensitizer.
Formula:
C₃₄H₅₀O₁₂ MW: 650.762 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00083511 CAS Number: 67526-95-8 |
Specification Test Results
Source/Host | Isolated from Thapsia garganica. |
Purity | >98% (HPLC) |
Appearance/Color | Colorless to white amorphous powder. |
Solubility | Soluble in DMSO or 100% ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...