Mouse Recombinant RANK (from HEK293 cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0092-C050 AG-40B-0092-3050
102515-592EA 332.31 USD
102515-592 102515-594
Mouse Recombinant RANK (from HEK293 cells)
Proteins and Peptides
RANK (TRANCE-R; CD265) is the receptor for RANKL/TRANCE/OPGL. It is essential for RANKL-mediated osteoclastogenesis and is involved in the regulation of interactions between T cells and dendritic cells.