Mouse Recombinant BCMA (from HEK293 cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0076-C050 AG-40B-0076-3050
102515-544EA 276.92 USD
102515-544 102515-546
Mouse Recombinant BCMA (from HEK293 cells)
Proteins and Peptides
BCMA is a receptor for BAFF and APRIL. It promotes B cell survival and plays a role in the regulation of humoral immunity. It's downstream signaling is dependent on NF-kappaB and JNK.