Human Recombinant TRAILR1 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
TRAIL-R1 (DR4; CD261) is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.