Mouse Recombinant Lipocalin 2 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0106-C010 AG-40A-0106-C050
102515-068EA 221.54 USD
102515-068 102515-070
Mouse Recombinant Lipocalin 2 (from HEK293 Cells)
Proteins and Peptides
Lipocalin-2 is an iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development.