Criterion TBE Precast Gels, Bio-Rad
Supplier: Bio-Rad
3450047 3450048 3450049
101645-812EA 29.23 USD
101645-812 101645-814 101645-818
Criterion TBE Precast Gels, Bio-Rad
Electrophoresis Gels Precast Gels
TBE gels are suitable for electrophoresis of nucleic acids from 50 to 2,000 base pairs; they are ideal for analysis of the purity of PCR products, standard dsDNA analysis, and RNase protection assays.
Criterion precast gels include a broad selection of 13.3 x 8.7 cm polyacrylamide gels in single-use cassettes. This gel size provides reproducible, high-resolution results with fast setup, loading, and run times.
Criterion gels are packaged and sold in individual units: 1–2 gels can be run in the Criterion cell system and 1–12 gels can be run in the high-throughput Criterion Dodeca cell system. The Criterion blotter offers a variety of convenient features, such as two electrode configurations and a convenient gel/blot assembly tray, for blotting applications.