Criterion™ XT Tris-Acetate Precast Gels, Midi Protein Gels, Bio-Rad
Supplier: Bio-Rad
3450130 3450129 3450131
101645-932EA 29.23 USD
101645-932 101645-930 101645-934
Criterion™ XT Tris-Acetate Precast Gels, Midi Protein Gels, Bio-Rad
Electrophoresis Gels Precast Gels
Criterion™ XT gels are ideal for vertical midi gel electrophoresis and are designed to work with optimized sample and running buffers without the need for antioxidant addition
  • Laemmli-type separation under denaturing conditions
  • Compatible with nonreducing and nondenaturing running and sample buffers
  • Neutral pH delays acrylamide hydrolysis significantly longer than Laemmli buffers
  • 8 month shelf life
  • Run 2-D gels in less than a day
Like traditional Laemmli systems, Criterion XT precast gels use discontinuous buffer ion fronts that form moving boundaries to stack and then separate proteins. Criterion XT Tris-acetate gels are formulated using a Tris-acetate buffer system (pH 7.0) that, when run with XT Tricine running buffer, separates large denatured proteins by molecular weight. For use with Criterion and Criterion Dodeca cells.
Información para pedidos: Criterion™ XT Tris-Acetate Precast Gels are packaged and sold in individual units: 1–2 gels can be run in Bio-Rad’s Criterion™ Cell, and 1–12 gels in the high-throughput Criterion™ Dodeca™ Cell.