Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: HC2001]

Supplier: Genscript
A02038-1 A02038-100
76510-092EA 3197.7 USD
76510-092 76510-094
Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: HC2001]
Antibodies
The product is specific for SARS-CoV-2 Spike Protein S1 subunit and its RBD domain.

  • COVID-19 antibodies
  • Validated for ELISA, WB, FACS and SARS-CoV-2 virus neutralization assay
  • Control antibodies and various antibody pairs available for ELISA-based detection
  • Neutralizing antibody standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards and control antibodies.

Type: Primary
Antigen: SARS-CoV-2 Spike S1
Clonality: Monoclonal
Clone: HC20More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR