Anti-Cetuximab Mouse Monoclonal Antibody [clone: 20H1]
Supplier: Genscript
A01939-40
76509-906EA 153.9 USD
76509-906
Anti-Cetuximab Mouse Monoclonal Antibody [clone: 20H1]
Antibodies
The product is specific for Cetuximab. This antibody can’t block Cetuximab binding with EGFR (Z03202, EGFR, His, Human) in ELISA. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A01938-40, Anti-Cetuximab Antibody (18D5), mAb, Mouse.
  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type: Primary
More Product Information