Anti-NTRK3 Mouse Monoclonal Antibody [clone: BS337]

Supplier: Biosensis
M-1837-100
76459-472EA 343.35 USD
76459-472
Anti-NTRK3 Mouse Monoclonal Antibody [clone: BS337]
Antibodies
Receptor tyrosine kinase involved in nervous system and probably heart development. Upon binding of its ligand NTF3/neurotrophin-3, NTRK3 autophosphorylates and activates different signaling pathways, including the phosphatidylinositol 3-kinase/AKT and the MAPK pathways, that control cell survival and differentiation (Ref: uniprot.org).

Applications: This antibody is recommended for use in flow cytometry (5-10 µg/mL) and immunocytochemistry (1-2 ug/mL). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NTRK3
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR