Anti-ARRS Mouse Monoclonal Antibody [clone: S128]
Supplier: Biosensis
M-1796-100
76334-010EA 343.35 USD
76334-010
Anti-ARRS Mouse Monoclonal Antibody [clone: S128]
Antibodies
Binds to photoactivated, phosphorylated RHO and terminates RHO signaling via G-proteins by competing with G-proteins for the same binding site on RHO (PubMed: 8003967, PubMed: 25205354). May play a role in preventing light-dependent degeneration of retinal photoreceptor cells (By similarity). Ref: uniprot.org
Antibody detects arrestin-1 protein only and does not cross-react with the other 3 arrestin molecules.
Applications: Western blotting (1:5,000) and Immunohistochemistry (1:1,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
More Product Information