Anti-IgG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: X63]

Supplier: Biosensis
M-1763-100-FT
76264-624EA 290.85 USD
76264-624
Anti-IgG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: X63]
Antibodies
Mouse monoclonal antibody (Clone X63) with no known antigen binding ability was purified from hybridoma cell culture medium by Protein G chromatography and labelled with FITC. X63-FITC is useful as a negative control antibody for immunofluorescence studies using FITC-labelled primary antibodies.

Immunofluorescence, flow cytometry. Useful as negative control antibody for FITC-labelled primary antibodies. Use at equal antibody concentration.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: X63
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR