IsoGel® Precast Agarose IEF Plates
Supplier: Lonza
56018 56015 56024
12002-194EA 840.27 USD
12002-194 12001-452 12002-192
IsoGel® Precast Agarose IEF Plates
Electrophoresis Gels Precast Gels
IsoGel® Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000 kDa.
  • Versatile - Fits most chambers
  • Convenient - No more pouring plates
The proteins can be quickly transferred from gel to membrane, stained in situ, or detected by antibodies within one hour of loading the gel. Each gel is supported on GelBond® film to provide dimensional stability throughout the IEF processing, and the convenient 125×100 mm (½×³/₈") gel size fits most horizontal IEF chambers. Each lot of plates is functionally tested and aMore Product Information