Anti-NTF3 Rabbit Polyclonal Antibody

Supplier: Biosensis
R-098-100
10782-388EA 343.35 USD
10782-388
Anti-NTF3 Rabbit Polyclonal Antibody
Antibodies
FUNCTION: Seems to promotes the survival of visceral and proprioceptive sensory neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family.

A cross reactivity of less than 1% to mouse NGF, recombinant human BDNF and 5% to NT4/5 has been shown by dot blot.
Application Information:
IHC, ELISA, WB, inhibition of biological activity. A dilution of 1:500 to 1:1000 is recommended for IHC, ELISA and western blot. For inhibition of biological activity: 1:10-50 for in vitro, 2-10 µl/g body weight for in vivo.More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol NTF3
Antigen Name Neurotrophin 3
Antigen Synonyms NGF-2|HDNF|Ntf3|Neurotrophin-3|Nerve growth factor 2|Ntf-3|Neurotrophic factor
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Chicken,Human,Rat
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Neutralisation Yes
Immunogen Recombinant human NT3
Format Lyophilized
Cross Adsorption 0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR