Anti-TGF-1 Chicken Polyclonal Antibody

Supplier: Biosensis
C-1541-100
10781-980EA 353.85 USD
10781-980
Anti-TGF-1 Chicken Polyclonal Antibody
Antibodies
Transforming growth factor beta-1 (TGFB1) is a multi-functional cytokine with roles in proliferation, differentiation and other functions in many cell types. The secreted TGFB1 protein is cleaved into a latency-associated peptide (LAP) and a mature TGFB1 peptide (Ref: SWISS-Prot).

Antibody recognizes TGFB1 by western blot and ELISAs; works best with cell supernatants or fluids as TGFB1 is secreted. Other applications have not been tested.
Application Information:
WB and ELISA. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determinedMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol TGFB1
Antigen Name Transforming Growth Factor Beta 1
Antigen Synonyms CED|TGFbeta|TGFB|DPD1|LAP
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Pig,Human,Rat,Mouse
Host Chicken
Isotype IgY
Western Blot Yes
ELISA Yes
Epitope Human TGFbeta-1 amino acids 372-386
Immunogen A peptide from human and mouse Transforming growth factor beta-1 (372-386 aa).
Format Liquid
Cross Adsorption 0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR