Anti-APMAP Chicken Polyclonal Antibody

Supplier: Biosensis
C-1524-100
10781-946EA 353.85 USD
10781-946
Anti-APMAP Chicken Polyclonal Antibody
Antibodies
Adiponectin is synthesized by adipocytes and is involved in the control of fat metabolism and insulin sensitivity, with direct anti-diabetic, anti-atherogenic and anti-inflammatory activities (ref: SWISSPROT).

This antibody detects adiponectin protein at approx 30 kDa in western blots from human, mouse and rat samples. Plasma makes a good positive control. The antibody has not been tested in IHC
Application Information:
WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: APMAP
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR