Human alpha1(I) Collagen (614-639) Peptide

Supplier: Anaspec Inc

AS-63837
103007-766EA 380 USD
103007-766
Human alpha1(I) Collagen (614-639) Peptide
Proteins and Peptides

This is a peptide inhibitor of collagen fibrillar matrix assembly.
Sequence:SAGFDFSFLPQPPQEKAHDGGRYYRA
MW:2942.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR