Human Lactoferricin B

Supplier: Anaspec Inc

AS-62651
103007-460EA 371.23 USD
103007-460
Human Lactoferricin B
Proteins and Peptides

This is amino acids 17 to 41 fragment of lactoferrin, known as lactoferricin B. This peptide exhibits anti-fungal properties in combination of other anti-fungal agents. Candida Albicans is one of the targets of the lactoferricin B.
Sequence:FKCRRWQWRMKKLGAPSITCVRRAF (S-S BOND)
MW:3123.9 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now

SPECIFICATIONS

Protein/Peptide Name Lactoferricin B
Protein Synonyms Lactoferricin Peptide|Lactoferrin Peptide fragment
Species Human
Conjugation Unconjugated
Sequence FKCRRWQWRMKKLGAPSITCVRRAF (S-S BOND)
Purity 95%
Molecular Weight 3123.9 Da
Storage Conditions –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR