HIV TAT-NSF222scr Fusion Polypeptide

Supplier: Anaspec Inc

AS-62210
103007-246EA 578.77 USD
103007-246
HIV TAT-NSF222scr Fusion Polypeptide
Proteins and Peptides

This is a scrambled TAT-NSF222scr fusion polypeptide. It is composed of 11 amino acids from the cell permeable human immunodeficiency virus TAT polypeptide, 3 glycines as a linker, followed by scrambled N-Ethyl-maleimide-sensitive factor (NSF) D1 domain. This peptide is used as a control for the TAT-NSF222 peptide.
Sequence: YGRKKRRQRRR-GGG-ENSFRFLADIFPAKAFPVRFE
MW: 4214.9 Da
% Peak area by HPLC: 95
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR