MAGE-3 (271-279)

Supplier: Anaspec Inc

AS-61355
103006-588EA 92.08 USD
103006-588
MAGE-3 (271-279)
Proteins and Peptides

This is a HLA-A*0201–restricted peptide derived from melanoma antigens encoded by MAGE-3.
Sequence:FLWGPRALV
MW:1058.3 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR