[Pyr1]-Apelin-13

Supplier: Anaspec Inc

AS-60833
103005-982EA 182.69 USD
103005-982
[Pyr1]-Apelin-13
Proteins and Peptides

[125I]-(Pyr1)Apelin-13 binds with high affinity and selectivity to human cardiac tissue.
Sequence:Pyr-RPRLSHKGPMPF-OH
MW:1533.8 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR