Leiurus quinquestriatus hebraeus Chlorotoxin (Cltx) 

Supplier: Anaspec Inc

AS-60770
103005-970EA 225.08 USD
103005-970
Leiurus quinquestriatus hebraeus Chlorotoxin (Cltx) 
Proteins and Peptides

A 36-amino acid Cl- channel blocker from Leiurus quinquestriatus scorpion venom.
Sequence:MCMPCFTTDHQMARKCDDCCGGKGRGKCYGPQCLCR-NH2 (Disulfide bridge: 2-19,5-28,16-33,20-35)
MW:3996 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR