Frog Sauvagine

Supplier: Anaspec Inc

AS-24475
103003-740EA 292.31 USD
103003-740
Frog Sauvagine
Proteins and Peptides

A hypotensive and diuretic peptide. Originally isolated from the skin of the frog, Phyllomedusa sauvagei. It affects diuresis in the cardiovascular system and causes the release of ACTH and endorphins.
Sequence:Pyr-GPPISIDLSLELLRKMIEIEKQEKEKQQAANNRLLLDTI-NH2
MW:4599.4 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR